missing translation for 'onlineSavingsMsg'
Learn More

4',7-Dihydroxyisoflavone, 97%

Product Code. 11449927
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11449927

Brand: Thermo Scientific Alfa Aesar B22877.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 486-66-8 | C15H10O4 | 254.241 g/mol

4',7-Dihydroxyisoflavone is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. It protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,7-Dihydroxyisoflavone is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. It protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

Solubility
Insoluble in water. Soluble in DMSO (25 mg/ml), ethanol (2 mg/ml), DMF (10 mg/ml), and 1:10 DMSO:PBS (pH 7.2) (0.15 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00016954
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Melting Point 328°C to 332°C (decomposition)
Quantity 5 g
Beilstein 231523
Merck Index 14,2801
Solubility Information Insoluble in water. Soluble in DMSO (25mg/ml),ethanol (2mg/ml),DMF (10mg/ml),and 1:10 DMSO:PBS (pH 7.2) (0.15mg/ml).
Formula Weight 254.24
Percent Purity 97%
Chemical Name or Material 4',7-Dihydroxyisoflavone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.