missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Pregnadien-6-methyl-17alpha-ol-3,20-dione Acetate-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Megestrol Acetate-D3 (17-acetate-D3) |
| Molecular Formula | C24 D3 H29 O4 |
| Quantity | 0.005 g |
| Synonym | 17-(Acetyl-2,2,2-d3-oxy)-6-methylpregna-4,6-diene-3,20-dione, Megestrol acetate-d3, Megestrol Acetate-D3 (17-acetate-D3), 17α-(Acetoxy-d3)-6-dehydro-6-methylprogesterone, 4,6-Pregnadien-6-methyl-17α-ol-3,20-dione Acetate-d3 |
| InChI Formula | InChI=1 S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h12-13,18-20 H,6-11H2,1-5H3/t18-,19+,20+,22-,23+,24+/m1/s1/i3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)O[C@@]1(CC[C@H]2 [C@@H]3 C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C(=O)C |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-17-yl] 2,2,2-trideuterioacetate |
| Molecular Weight (g/mol) | 387.527 |
| Formula Weight | 387.2489 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction