Learn More
4,5-Dihydro Prednisone, TRC
Specifications
Specifications
| Chemical Name or Material | 4,5-Dihydro Prednisone |
| CAS | 103881-93-2 |
| Molecular Formula | C21 H28 O5 |
| Quantity | 1 mg |
| Synonym | 4,5-Dihydro Prednisone, Pregn-1-ene-3,11,20-trione, 17,21-dihydroxy-, (5α)-, 5α-Pregn-1-ene-3,11,20-trione, 17,21-dihydroxy- (6CI), (5α)-17,21-Dihydroxypregn-1-ene-3,11,20-trione |
| InChI Formula | InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,12,14-15,18,22,26H,3-4,6,8-11H2,1-2H3/t12-,14-,15-,18+,19-,20-,21-/m0/s1 |
| SMILES | C[C@]12CC(=O)[C@H]3[C@@H](CC[C@H]4CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO |
| IUPAC Name | (5S,8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-4,5,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione |
| Molecular Weight (g/mol) | 360.444 |
| Formula Weight | 360.194 |