Learn More
4,5-Difluoro-2-methoxybenzyl bromide, 97%, Thermo Scientific™
Product Code. 11346839
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346839 5 g 5g
11336839 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11346839 Supplier Thermo Scientific Alfa Aesar Supplier No. H26764.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 886499-64-5
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.044
MDL Number MFCD06660220
InChI Key PRLKVWCFDNQAED-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-methoxybenzyl bromide, 1-bromomethyl-4,5-difluoro-2-methoxybenzene
PubChem CID 17750791
IUPAC Name 1-(bromomethyl)-4,5-difluoro-2-methoxybenzene
SMILES COC1=CC(=C(C=C1CBr)F)F
Specifications
Melting Point 40°C to 43°C
Quantity 5 g
UN Number UN3261
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 4,5-Difluoro-2-methoxybenzyl bromide
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.