Learn More
4',5'-Bis(1,3,2-dithiarsolan-2-yl)-3',6'-dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthene]-6-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4',5'-Bis(1,3,2-dithiarsolan-2-yl)-3',6'-dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthene]-6-carboxylic Acid |
| CAS | 1042084-20-7 |
| Molecular Formula | C25 H18 As2 O7 S4 |
| Quantity | 5 mg |
| Synonym | 4',5'-di-1,3,2-Dithiarsolan-2-yl-3',6'-dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthene]-6-carboxylic Acid, 6-CrAsH-EDT2 |
| InChI Formula | InChI=1S/C25H18As2O7S4/c28-17-5-3-14-21(19(17)26-35-7-8-36-26)33-22-15(4-6-18(29)20(22)27-37-9-10-38-27)25(14)16-11-12(23(30)31)1-2-13(16)24(32)34-25/h1-6,11,28-29H,7-10H2,(H,30,31) |
| SMILES | OC(=O)c1ccc2C(=O)OC3(c4ccc(O)c([As]5SCCS5)c4Oc6c([As]7SCCS7)c(O)ccc36)c2c1 |
| IUPAC Name | 4',5'-bis(1,3,2-dithiarsolan-2-yl)-3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid |
| Molecular Weight (g/mol) | 708.5094 |
| Formula Weight | 707.8367 |