Learn More
4,4'-Thiobis(6-tert-butyl-o-cresol), TRC
Specifications
Specifications
| Chemical Name or Material | 4,4'-Thiobis(6-tert-butyl-o-cresol) |
| CAS | 96-66-2 |
| Molecular Formula | C22H30O2S |
| Quantity | 5 g |
| Synonym | Bis(3-tert-butyl-4-hydroxy-5-methylphenyl) Sulfide, 2,2'-Di-tert-Butyl-6,6'-dimethyl-4,4'-thiodiphenol, 2-tert-Butyl-4-(3-tert-butyl-4-hydroxy-5-methylphenyl)sulfanyl-6-methylphenol, 4,4'-Thiobis(2-methyl-6-tert-butylphenol), 4,4'-Thiobis(2-tert-butyl-6-methylphenol), 4,4'-Thiobis(6-tert-butyl-2-methylphenol), 4,4'-Thiobis(6-tert-butyl-o-cresol), 4,4'-Thiobis-(6-tert-butyl-2-methylphenol), AO 736, Antioxidant 736, Antioxidant E 736, E 736, Ethyl 736, Ethyl AO 736, Ethyl Antioxidant 736, KY 736, NSC 58409, TB 2, TB 2 (antioxidant) |
| InChI Formula | InChI=1S/C22H30O2S/c1-13-9-15(11-17(19(13)23)21(3,4)5)25-16-10-14(2)20(24)18(12-16)22(6,7)8/h9-12,23-24H,1-8H3 |
| SMILES | Cc1cc(Sc2cc(C)c(O)c(c2)C(C)(C)C)cc(c1O)C(C)(C)C |
| IUPAC Name | 4,4'-thiobis(2-(tert-butyl)-6-methylphenol) |
| Molecular Weight (g/mol) | 358.24 |
| Formula Weight | 358.1967 |