Learn More
4-(4-Methoxyphenoxy)-2-methyl-1-(4-methylbenzenesulfonate) 1,2-Butanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(4-Methoxyphenoxy)-2-methyl-1-(4-methylbenzenesulfonate) 1,2-Butanediol |
| CAS | 303765-54-0 |
| Molecular Formula | C18 H31 N5 O6 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C18H31N5O6/c19-11(17(28)29)5-2-4-8-23-10-14(25)13(24)9-12-16(23)22-18(21-12)20-7-3-1-6-15(26)27/h11-14,24-25H,1-10,19H2,(H,20,21)(H,26,27)(H,28,29)/t11-,12-,13-,14+/m0/s1 |
| SMILES | N[C@@H](CCCCN1C[C@@H](O)[C@@H](O)C[C@@H]2N=C(NCCCCC(=O)O)N=C12)C(=O)O |
| IUPAC Name | (2S)-6-[(6R,7S,8aS)-2-(4-carboxybutylamino)-6,7-dihydroxy-6,7,8,8a-tetrahydro-5H-imidazo[4,5-b]azepin-4-yl]-2-aminohexanoic acid |
| Molecular Weight (g/mol) | 413.469 |
| Formula Weight | 413.227 |