Learn More
4-(4-Hydroxyphenoxy)benzoic acid, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 500-76-5 |
| Molecular Formula | C13H10O4 |
| MDL Number | MFCD00059645 |
| Quantity | 1 g |
| SMILES | O=C(O)C1=CC=C(OC2=CC=C(O)C=C2)C=C1 |
| Molecular Weight (g/mol) | 230.22 |
| Percent Purity | 96% |
| Chemical Name or Material | 4-(4-Hydroxyphenoxy)benzoic acid |