Learn More
4-(4-Fluorophenyl)-4-oxobutanoic Acid, TRC
Product Code. 30402414
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402414 5 g -
30398545 2.5 g 2.5g
30401023 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30402414 Supplier Toronto Research Chemicals Supplier No. TRCF6322835G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 366-77-8
Molecular Formula C10 H9 F O3
Molecular Weight (g/mol) 196.175
Synonym Benzenebutanoic acid, 4-fluoro-γ-oxo- (9CI, ACI), 4-Fluoro-γ-oxobenzenebutanoic acid (ACI), Propionic acid, 3-(p-fluorobenzoyl)- (7CI, 8CI), 3-(4-Fluorobenzoyl)propanoic acid, 3-(4-Fluorobenzoyl)propionic acid, 3-(p-Fluorobenzoyl)propionic acid, 4-(4-Fluorophenyl)-4-oxobutanoic acid, 4-(4-Fluorophenyl)-4-oxobutyric acid, 4-Fluorobenzoylpropionic acid, 4-Oxo-4-(p-fluorophenyl)butyric acid, Ba 2833, NSC 408180, β-(4-Fluorobenzoyl)propionic acid, β-(p-Fluorobenzoyl)propionic acid, γ-(4-Fluorophenyl)-γ-oxobutanoic acid
IUPAC Name 4-(4-fluorophenyl)-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)c1ccc(F)cc1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H9FO3/c11-8-3-1-7(2-4-8)9(12)5-6-10(13)14/h1-4H,5-6H2,(H,13,14)
Formula Weight 196.054
Chemical Name or Material 4-(4-Fluorophenyl)-4-oxobutanoic Acid