Learn More
4,4'-Dihydroxydiphenyl Ether, TRC
Product Code. 30373343
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373343 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30373343 Supplier Toronto Research Chemicals Supplier No. TRCD493435500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4,4'-Dihydroxydiphenyl Ether
CAS 1965-09-9
Purity Grade Notes HPLC
Molecular Formula C12H10O3
Quantity 500 mg
InChI Formula InChI=1S/C12H10O3/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8,13-14H
SMILES Oc1ccc(Oc2ccc(O)cc2)cc1
Molecular Weight (g/mol) 202.21
Formula Weight 202.063
Percent Purity >95