Learn More
4,4'-Dibromobiphenyl-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-bromo-4-(4-bromo-2,3,5,6-tetradeuteriophenyl)-2,3,5,6-tetradeuteriobenzene |
| CAS | 80523-79-1 |
| Molecular Formula | C12 2H8 Br2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C12H8Br2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Br)c2c([2 H])c([2 H])c(Br)c([2 H])c2[2 H] |
| IUPAC Name | 1-bromo-4-(4-bromo-2,3,5,6-tetradeuteriophenyl)-2,3,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 320.049 |
| Formula Weight | 317.949 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |