Learn More
4-(4'-Chloro-4-biphenylylsulfonylamino)benzoic acid, 96%, Thermo Scientific™
Product Code. 15418975
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418975 250 mg 250mg
15408975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15418975 Supplier Thermo Scientific Alfa Aesar Supplier No. H34497.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 885269-32-9
Molecular Formula C19H14ClNO4S
Molecular Weight (g/mol) 387.834
MDL Number MFCD06409447
InChI Key AYWSYJTWHIPSFL-UHFFFAOYSA-N
Synonym 4-4'-chloro-4-biphenylylsulfonylamino benzoic acid, 4-4'-chloro-1,1'-biphenyl-4-sulfonamido benzoic acid, 4-4'-chloro-1,1'-biphenyl-4-ylsulfonamido benzoic acid
PubChem CID 44669160
IUPAC Name 4-[[4-(4-chlorophenyl)phenyl]sulfonylamino]benzoic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)NC3=CC=C(C=C3)C(=O)O
Specifications
Melting Point 298°C
Quantity 250 mg
Formula Weight 387.84
Percent Purity 96%
Chemical Name or Material 4-(4'-Chloro-4-biphenylylsulfonylamino)benzoic acid