Learn More
4-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic Acid, TRC
Product Code. 30405833
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405833 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30405833 Supplier Toronto Research Chemicals Supplier No. TRCF056450500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic Acid
CAS 1017384-80-3
Molecular Formula C16 H17 F N2 O4
Quantity 500 mg
InChI Formula InChI=1S/C16H17FN2O4/c17-11-1-2-12-13(9-11)23-18-16(12)10-5-7-19(8-6-10)14(20)3-4-15(21)22/h1-2,9-10H,3-8H2,(H,21,22)
SMILES OC(=O)CCC(=O)N1CCC(CC1)c2noc3cc(F)ccc23
IUPAC Name 4-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic acid
Molecular Weight (g/mol) 320.316
Formula Weight 320.117