missing translation for 'onlineSavingsMsg'
Learn More

4-[4-[2-(2-Pyridinyloxy)propoxy]phenoxy]phenol, TRC

Product Code. 30136709
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136709 100 mg 100mg
30134856 50 mg 50mg
30012479 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136709 Supplier Toronto Research Chemicals Supplier No. TRCP840590100MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159600-61-0
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.37
Synonym Pyriproxyfen-4'-hydroxy, Phenol, 4-[4-[2-(2-pyridinyloxy)propoxy]phenoxy]- (9CI, ACI), 4-[4-[2-(2-Pyridinyloxy)propoxy]phenoxy]phenol
IUPAC Name 4-[4-(2-pyridin-2-yloxypropoxy)phenoxy]phenol
SMILES CC(COc1ccc(Oc2ccc(O)cc2)cc1)Oc3ccccn3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H19NO4/c1-15(24-20-4-2-3-13-21-20)14-23-17-9-11-19(12-10-17)25-18-7-5-16(22)6-8-18/h2-13,15,22H,14H2,1H3
Formula Weight 337.1314
Chemical Name or Material 4-[4-[2-(2-Pyridinyloxy)propoxy]phenoxy]phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.