Learn More
4,4'-(1-Methylene) bis-Benzonitrile, TRC
Product Code. 30483111
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483111 50 mg 50mg
30479781 10 mg 10mg
30476733 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30483111 Supplier Toronto Research Chemicals Supplier No. TRCM22144050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10466-37-2
Molecular Formula C15H10N2
Molecular Weight (g/mol) 218.25
Synonym Benzonitrile, 4,4'-methylenebis-, Benzonitrile, 4,4'-methylenedi- (6CI,7CI,8CI), 4,4'-Methylenebis[benzonitrile], 4,4'-Dicyanodiphenylmethane, NSC 174860, 4,4'-Methylenedibenzonitrile
IUPAC Name 4-[(4-cyanophenyl)methyl]benzonitrile
SMILES N#Cc1ccc(Cc2ccc(cc2)C#N)cc1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C15H10N2/c16-10-14-5-1-12(2-6-14)9-13-3-7-15(11-17)8-4-13/h1-8H,9H2
Formula Weight 218.0844
Chemical Name or Material 4,4'-(1-Methylene) bis-Benzonitrile