missing translation for 'onlineSavingsMsg'
Learn More

4-(3,4-Dimethoxyphenyl)butanoic Acid, TRC

Product Code. 30370680
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370680 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370680 Supplier Toronto Research Chemicals Supplier No. TRCD4610901G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(3,4-Dimethoxyphenyl)butanoic Acid
CAS 13575-74-1
Molecular Formula C12H16O4
Quantity 1 g
Synonym 3,4-Dimethoxybenzenebutanoic Acid, 4-(3,4-Dimethoxyphenyl)butyric Acid, 3,4-Dimethoxybenzenebutanoic Acid, 4-(3,4-Dimethoxyphenyl)butanoic Acid, 4-(3,4-Dimethoxyphenyl)butyric Acid, NSC 286140
InChI Formula InChI=1S/C12H16O4/c1-15-10-7-6-9(8-11(10)16-2)4-3-5-12(13)14/h6-8H,3-5H2,1-2H3,(H,13,14)
SMILES COc1ccc(CCCC(=O)O)cc1OC
Molecular Weight (g/mol) 224.25
Formula Weight 224.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.