Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Vo49
Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Description
- 4 to 20 mA current loop interface
- Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specifications
Specifications
| For Use With (Equipment) | Models 30, 55, 75, 110, 160, 260, 450, 750, 1060 |
| Type | Current Loop Interface |
| Amperage | 4 to 20 mA |