missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)benzoic acid, 96%

Product Code. 15456487
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15456487

Brand: Thermo Scientific Alfa Aesar H33642.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29886-62-2 | C11H8O2S | 204.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29886-62-2
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
MDL Number MFCD00690742
InChI Key CVDUBQJEQNRCIZ-UHFFFAOYSA-N
Synonym 4-thiophen-2-yl benzoic acid, 4-2-thienyl benzoic acid, 4-thiophen-2-yl-benzoic acid, benzoic acid,4-2-thienyl, benzoic acid, 4-2-thienyl, pubchem12353, acmc-1cb4z, 4-2-thienyl benzoicacid, 4-thien-2-ylbenzoic acid, amtda058
PubChem CID 736498
IUPAC Name 4-thiophen-2-ylbenzoic acid
SMILES C1=CSC(=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 243°C to 245°C
Quantity 5 g
Formula Weight 204.25
Percent Purity 96%
Chemical Name or Material 4-(2-Thienyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.