Learn More
4’,2’’-O-Di(trimethysilyl)tylosin, TRC
Specifications
Specifications
| Chemical Name or Material | 4’,2’’-O-Di(trimethysilyl)tylosin |
| CAS | 114442-77-2 |
| Molecular Formula | C52H93NO17Si2 |
| Quantity | 10 mg |
| Synonym | 2A,4C-Bis-O-(trimethylsilyl)-tylosin |
| InChI Formula | InChI=1S/C52H93NO17Si2/c1-19-39-36(28-62-50-48(61-12)47(60-11)45(33(6)64-50)69-71(13,14)15)24-29(2)20-21-37(55)30(3)25-35(22-23-54)43(31(4)38(56)26-40(57)66-39)68-51-46(70-72(16,17)18)42(53(9)10)44(32(5)65-51)67-41-27-52(8,59)49(58)34(7)63-41/h20-21,23-24,30-36,38-39,41-51,56,58-59H,19,22,25-28H2,1-18H3/b21-20+,29-24+/t30?,31?,32?,33?,34?,35-,36?,38+,39+,41-,42-,43+,44+,45?,46?,47?,48?,49-,50+,51-,52?/m0/s1 |
| SMILES | O=C(C[C@@H](O)[C@H](C)[C@@H](O[C@H]1[C@H](O[Si](C)(C)C)[C@@H](N(C)C)[C@H](O[C@@H]2O[C@@H](C)[C@H](O)[C@@](O)(C)C2)[C@@H](C)O1)[C@@H](CC=O)C[C@H]3C)O[C@H](CC)C(CO[C@@H]4O[C@H](C)[C@@H](O[Si](C)(C)C)[C@@H](OC)[C@H]4OC)/C=C(C)/C=C/C3=O |
| IUPAC Name | 2-((4R,5S,6S,7R,9R,11E,13E,16R)-6-(((2R,3R,4S,5S,6R)-5-(((2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-(dimethylamino)-6-methyl-3-((trimethylsilyl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-15-((((2R,3R,4S,5R,6R)-3,4-dimethoxy-6-methyl-5-((trimethylsilyl)oxy)tetrahydro-2H-pyran-2-yl)oxy)methyl)-16-ethyl-4-hydroxy-5,9,13-trimethyl-2,10-dioxooxacyclohexadeca-11,13-dien-7-yl)acetaldehyde |
| Molecular Weight (g/mol) | 1060.46 |
| Formula Weight | 1059.6 |