missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyphenyl)-2-(1-piperidinyl)thiazole, 97%, Thermo Scientific™

Product Code. 15464738
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15464738

Brand: Thermo Scientific Alfa Aesar H58462.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916497-18-2
Molecular Formula C14H16N2OS
Molecular Weight (g/mol) 260.36
MDL Number MFCD22682977
InChI Key CIZFLLFRDXZCLM-UHFFFAOYSA-N
Synonym 2-2-piperidin-1-yl-1,3-thiazol-4-yl phenol, 4-2-hydroxyphenyl-2-1-piperidinyl thiazole
PubChem CID 15984509
IUPAC Name 2-(2-piperidin-1-yl-1,3-thiazol-4-yl)phenol
SMILES OC1=CC=CC=C1C1=CSC(=N1)N1CCCCC1

Specifications

Quantity 1 g
Formula Weight 289.29
Percent Purity 97%
Chemical Name or Material 4-(2-Hydroxyphenyl)-2-(1-piperidinyl)thiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.