Learn More
4-(2-Benzoyl-4-chlorophenyl)-5-methyl-4H-1,2,4-triazole-3-carboxaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2-Benzoyl-4-chlorophenyl)-5-methyl-4H-1,2,4-triazole-3-carboxaldehyde |
| CAS | 36916-20-8 |
| Molecular Formula | C17H12ClN3O2 |
| Quantity | 10 mg |
| Synonym | 5-Chloro-2-(3-formyl-5-methyl-4H-1,2,4-triazol-4-yl)benzophenone |
| InChI Formula | InChI=1S/C17H12ClN3O2/c1-11-19-20-16(10-22)21(11)15-8-7-13(18)9-14(15)17(23)12-5-3-2-4-6-12/h2-10H,1H3 |
| SMILES | CC1=NN=C(C=O)N1C2=C(C=C(C=C2)Cl)C(=O)C3=CC=CC=C3 |
| IUPAC Name | 4-(2-benzoyl-4-chlorophenyl)-5-methyl-4H-1,2,4-triazole-3-carbaldehyde |
| Molecular Weight (g/mol) | 325.75 |
| Formula Weight | 325.06 |