missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoanilino)pyridine, TRC

Product Code. 30327117
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327117 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327117 Supplier Toronto Research Chemicals Supplier No. TRCA58316025MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(2-Aminoanilino)pyridine
CAS 65053-26-1
Molecular Formula C11 H11 N3
Quantity 25 mg
Synonym Droperidol Impurity 5
InChI Formula InChI=1S/C11H11N3/c12-10-3-1-2-4-11(10)14-9-5-7-13-8-6-9/h1-8H,12H2,(H,13,14)
SMILES Nc1ccccc1Nc2ccncc2
IUPAC Name 2-N-pyridin-4-ylbenzene-1,2-diamine
Molecular Weight (g/mol) 185.23
Formula Weight 185.0953
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.