missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)(nitroso)amino)benzoyl-2,3,5,6-d4)glutamic Acid, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | (4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)(nitroso)amino)benzoyl-2,3,5,6-d4)glutamic Acid |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 D4 H14 N8 O7 |
| Quantity | 10 mg |
| Synonym | Nitrosofolic Acid-D4 |
| InChI Formula | InChI=1S/C19H18N8O7/c20-19-24-15-14(17(31)25-19)22-10(7-21-15)8-27(26-34)11-3-1-9(2-4-11)16(30)23-12(18(32)33)5-6-13(28)29/h1-4,7,12H,5-6,8H2,(H,23,30)(H,28,29)(H,32,33)(H3,20,21,24,25,31)/t12-/m0/s1/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c(C(=O)N[C@@H](CCC(=O)O)C(=O)O)c([2H])c([2H])c1N(Cc2cnc3N=C(N)NC(=O)c3n2)N=O |
| IUPAC Name | (2S)-2-[[4-[(2-amino-4-oxo-3H-pteridin-6-yl)methyl-nitrosoamino]-2,3,5,6-tetradeuteriobenzoyl]amino]pentanedioic acid |
| Molecular Weight (g/mol) | 474.42 |
| Formula Weight | 474.155 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction