missing translation for 'onlineSavingsMsg'
Learn More

4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid, 98+%

Product Code. p-7036563
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461706 1 g 1g
15471706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461706 Supplier Thermo Scientific Alfa Aesar Supplier No. H54874.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 126828-35-1 | C32H29NO7 | 539.584 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126828-35-1
Molecular Formula C32H29NO7
Molecular Weight (g/mol) 539.584
MDL Number MFCD00153509
InChI Key UPMGJEMWPQOACJ-UHFFFAOYSA-N
Synonym rink amide linker, 4-2,4-dimethoxyphenyl fmoc-amino methyl phenoxyacetic acid, 2-4-9h-fluoren-9-yl methoxy carbonyl amino 2,4-dimethoxyphenyl methyl phenoxy acetic acid, rink amide linker;, fmoc-knorr, knorr linker;, fmoc-knorr-linker, ambotzrl-1027, pubchem12817, acmc-20amr2
PubChem CID 2761459
IUPAC Name 2-[4-[(2,4-dimethoxyphenyl)-(9H-fluoren-9-ylmethoxycarbonylamino)methyl]phenoxy]acetic acid
SMILES COC1=CC(=C(C=C1)C(C2=CC=C(C=C2)OCC(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)OC

Specifications

Melting Point ∽180°C (decomposition)
Quantity 1 g
Formula Weight 539.58
Percent Purity ≥98%
Chemical Name or Material 4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.