missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 15440506
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15440506

Brand: Thermo Scientific Alfa Aesar H53245.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-31-9
Molecular Formula C8H9BClN5O3
Molecular Weight (g/mol) 269.45
MDL Number MFCD06659882
InChI Key AZODLVUFJXZNTC-UHFFFAOYSA-N
Synonym 4-1h-tetrazol-5-yl carbamoyl phenyl boronic acid hydrochloride, 4-1h-tetrazol-5-ylcarbamoyl benzeneboronic acid hydrochloride, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid, hcl, 4-1h-tetrazol-5-yl-carbamoyl benzeneboronic acid, hcl, 4-1h-1,2,3,4-tetrazol-5-yl carbamoyl phenylboronic acid hydrochloride, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hcl, 4-1h-tetrazol-5-yl-carbamoyl phenylboronic acid hydrochloride, 4-1h-tetrazol-5-yl carbamoyl phenyl boronicacidhydrochloride, 4-1h-tetrazol-5-ylaminocarbonyl phenylboronic acid hydrochloride, 4-2h-tetrazol-5-yl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
PubChem CID 44119407
IUPAC Name [4-(2H-tetrazol-5-ylcarbamoyl)phenyl]boronic acid;hydrochloride
SMILES Cl.OB(O)C1=CC=C(C=C1)C(=O)NC1=NNN=N1

Specifications

Melting Point >300°C
Quantity 250 mg
Formula Weight 269.45
Percent Purity 97%
Chemical Name or Material 4-(1H-Tetrazol-5-ylcarbamoyl)benzeneboronic acid hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.