missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-pyrazol-1-ylmethyl)benzaldehyde, Thermo Scientific™

Product Code. 10506985
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10506985 5 g 5g
10483034 250 mg 250mg
10637123 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10506985 Supplier Thermo Scientific Maybridge Supplier No. CC56004DE
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 887922-90-9
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.21
MDL Number MFCD08059835
InChI Key FAURNROAEFQBHX-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl benzaldehyde, 4-pyrazol-1-ylmethyl-benzaldehyde, 4-1h-pyrazol-1-yl methyl benzaldehyde, 4-pyrazol-1-ylmethyl benzaldehyde, 4-pyrazolylmethyl benzaldehyde, 4-1h-pyrazol-1-ylmethyl-benzaldehyde, benzaldehyde,4-1h-pyrazol-1-ylmethyl
PubChem CID 18525876
IUPAC Name 4-(pyrazol-1-ylmethyl)benzaldehyde
SMILES O=CC1=CC=C(CN2C=CC=N2)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.