Learn More
4-(1-Pyrrolidinylsulforylmenthyl)phenylhydrazine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(1-Pyrrolidinylsulforylmenthyl)phenylhydrazine Hydrochloride |
| CAS | 334981-11-2 |
| Molecular Formula | C11H18ClN3O2S |
| Quantity | 1 g |
| Synonym | 1-[[(4-Hydrazinophenyl)methyl]sulfonyl]pyrrolidine Hydrochloride, 1-((4-(Hydrazino)benzenemethane)sulfonyl)pyrrolidine Hydrochloride |
| InChI Formula | InChI=1S/C11H17N3O2S.ClH/c12-13-11-5-3-10(4-6-11)9-17(15,16)14-7-1-2-8-14;/h3-6,13H,1-2,7-9,12H2;1H |
| SMILES | Cl.NNc1ccc(CS(=O)(=O)N2CCCC2)cc1 |
| IUPAC Name | [4-(pyrrolidin-1-ylsulfonylmethyl)phenyl]hydrazine;hydrochloride |
| Molecular Weight (g/mol) | 291.8 |
| Formula Weight | 291.0808 |