missing translation for 'onlineSavingsMsg'
Learn More

(3S)-3-Hydroxy Quinidine, TRC

Product Code. 30395694
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395694 1 mg
30404013 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395694 Supplier Toronto Research Chemicals Supplier No. TRCH9532301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53467-23-5
Molecular Formula C20 H24 N2 O3
Molecular Weight (g/mol) 340.42
IUPAC Name (2R,4S,5S)-5-ethenyl-2-[(S)-hydroxy-(6-methoxyquinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-5-ol
SMILES COc1ccc2nccc([C@H](O)[C@H]3C[C@@H]4CCN3C[C@]4(O)C=C)c2c1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H24N2O3/c1-3-20(24)12-22-9-7-13(20)10-18(22)19(23)15-6-8-21-17-5-4-14(25-2)11-16(15)17/h3-6,8,11,13,18-19,23-24H,1,7,9-10,12H2,2H3/t13-,18+,19-,20+/m0/s1
Formula Weight 340.1787
Percent Purity >95%
Chemical Name or Material (3S)-3-Hydroxy Quinidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.