Learn More
(3R,5R)-Atorvastatin Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (3R,5R)-Atorvastatin Sodium Salt |
| CAS | 134523-01-6 |
| Molecular Formula | C33 H34 F N2 O5 . Na |
| Quantity | 25 mg |
| Synonym | (3R,5R)-Atorvastatin Sodium Salt, (βR,δR)-2-(4-Fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic Acid Monosodium Salt, [R-(R*,R*)]-2-(4-Fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic Acid Monosodium Salt |
| InChI Formula | InChI=1S/C33H35FN2O5.Na/c1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40;/h3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);/q;+1/p-1/t26-,27-;/m1./s1 |
| SMILES | [Na+].CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)[O-] |
| IUPAC Name | sodium;(3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate |
| Molecular Weight (g/mol) | 580.622 |
| Formula Weight | 580.235 |