Learn More
Enzymaster (3R,5R)-3,5-Heptanediol, >99%
Product Code. 17804651
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17804651 5 g 5g
17884641 10 g 10g
17894641 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17804651 Supplier Enzymaster Supplier No. 10415
(3R,5R)-3,5-Heptanediol with very high chemical and optical purities >99%.
  • Enzymatically produced
  • Chemical purity >99%
  • Enantiomeric excess >99%
  • Diastereomeric excess >99%
Specifications
Melting Point 52°C
Color White
Boiling Point 222°C to 238°C
Enantiomeric Excess >99%
Quantity 5 g
Molecular Weight (g/mol) 132.2 g/mol
Percent Purity >99%
Grade Reagent
Physical Form Solid
Diastereomeric Excess >99%