Learn More
3M™ Versaflo™ Uncoated Visor for M-Series PAPR
Product Code. 15779727 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-series PAPR
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15779727 M-series PAPR Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15779727 Supplier 3M™ Supplier No. 7000042917
  • ONLINE EXCLUSIVE PRICE
    254.00€
    184.42€ / Pack of 5
    Save 69.58€
    27% Off
View all promotions
Estimated Shipment: 09-12-2026
Provides additional molten metal splash protection for M-Series Headtops
  • Provides additional molten metal splash protection for M-Series Headtops
  • 3M Catalog# M-925
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) M-series PAPR
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
Includes One m-921 visor gasket
Material Polycarbonate
Model M-925
Product Type Standard Visor