Learn More
3M™ Versaflo™ M-100 Series PAPR Replacement Head Suspension
Product Code. 11707424 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-100 Faceshields
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11707424 M-100 Faceshields Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11707424 Supplier 3M™ Supplier No. 7100063886
  • ONLINE EXCLUSIVE PRICE
    95.00€
    64.16€ / Pack of 1
    Save 30.84€
    32% Off
Estimated Shipment: 14-01-2027
Head suspension is a replacement part for Versaflo™ M-100 Series faceshields.
  • 3M Catalog# M-150
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Reusable
Product Type Head Suspension
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) M-100 Faceshields
Model M-150