Learn More
3M™ TR-300 and TR-600 PAPR Battery Packs
Product Code. p-57862
Click to view available options
For Use With (Equipment):
Versaflo TR-300 PAPR Series
Versaflo TR-600 PAPR Series
Battery Life:
4 to 6 hr.
8 to 12 hr.
8 to 18 hr.
Color:
Black
Blue, Gray
Unit Size:
Each
Product Code. 11793896
Brand: 3M™ 7100006660
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Choose from a selection of lithium ion rechargeable batteries for the TR-300 or TR-600 PAPR systems.
  • Rechargeable battery packs
  • Lithium Ion
  • Long run time for filters and cartridges
Battery packs must be used in accordance with the user instructions, warnings and limitations accompanying each product
Compliance
NIOSH approved.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 4 to 6 hr.
Battery Type Lithium-ion
Color Black
Product Type Battery Pack
For Use With (Application) Supplying power for PAPR systems
For Use With (Equipment) Versaflo TR-300 PAPR Series
Model TR-330
Recharge Time <3 hr.
Weight (English) 2 lb.
Weight (Metric) 0.918 kg