Learn More
3M™ Full Facepiece Respirator 7800S Parts and Accessories
Product Code. 15789767
Click to view available options
Product Type:
Exhalation Valve
Inhalation Valve
Unit Size:
Pack of 10
Pack of 20
Product Code. 15789767
Brand: 3M™ 7000051927
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with 3M™ 7000 Series Half- and Full-Facepiece Respirators
  • Replacement parts extend the life of the respirator
  • Accessories increase versatility
  • 3M Catalog# 7282, 7283, 7884, 7893S, 7894
TRUSTED_SUSTAINABILITY
Specifications
Product Type Exhalation Valve
For Use With (Application) Grinding, welding
For Use With (Equipment) 7800S Full Facepiece Respirator
Model 7283
Dimensions (L x W x H) 7.4 x 6.4 x 4 in. (18.7 x 16.2 x 10.1 cm)
Length (English) 7.4 in.
Length (Metric) 18.7 cm
Width (English) 6.4 in.
Width (Metric) 16.2 cm
Height (English) 4 in.
Height (Metric) 10.1 cm
Weight (English) 0.75 lb.
Weight (Metric) 0.34 kg