Learn More
3M™ Accessories and Replacement Parts for Full-Facepiece Respirators, Lens assembly
Product Code. 15759637 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
6000 series full facepiece respirator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15759637 6000 series full facepiece respirator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15759637 Supplier 3M™ Supplier No. 7000002113
  • ONLINE EXCLUSIVE PRICE
    106.00€
    72.84€ / Pack of 1
    Save 33.16€
    31% Off
Estimated Shipment: 10-12-2026
Lens assembly for use with full-facepiece respirators.
  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
  • 3M Catalog# 6898
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH
Color Clear
Product Type Lens Assembly
For Use With (Application) Automotive, metal production, fabrication, pharmaceutical, transportation, military maintenance, construction, food and beverage manufacturing
For Use With (Equipment) 6000 series full facepiece respirator
Includes Inhalation port fitting, inhalation valves and inhalation port filter/cartridge gaskets
Model 6898