Learn More
Corning™ 3D Clear Starter Kit
Shop All Corning Products
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 5
Description
- Rapid tissue clearing
- Easy to Use
- No Special Equipment Required
- Compatible with IF, FP and other Fluorescent labels
- Microplate and Automation Compatible
- Reversible to Follow up with 2D H&E/IHC
Specifications
Specifications
| Quantity | 1 Kit |
| Includes | 3 D Clear Reagent, Antibody, Blocking, Penetration, and Washing Buffers |