Learn More
Greiner Bio-One 384-well F-Bottom Polystyrene Microplates
Click to view available options
Color:
Black
White
Sterility:
Non-sterile
Sterile
Surface Treatment:
FLUOTRAC 200
FLUOTRAC 600
LUMITRAC 200
LUMITRAC 600
Unit Size:
Pack of 40
Description
- Non-Pyrogenic
- Material: Polystyrene
- Free of detectable DNase, RNase, human DNA
- Traced way back to production through defined LOT number system
- Well-to-well spacing: 4.5mm
Certifications
DIN ISO 9001
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Binding Type | High Binding |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | LUMITRAC 600 |
| Well Shape | Flat Bottom (F-well) |