Learn More
Greiner Bio-One 384-well μClear™ Bottom Polystyrene Microplates
missing translation for 'orderingAttributeHoverText'
Color:
Clear
White, Clear Bottom
Sterility:
Sterile
Binding Property:
High Binding
missing translation for 'unitSize'
Pack of 40
Description
- Non-Pyrogenic
- Material: Polystyrene
- Free of detectable DNase, RNase, human DNA
- Traced way back to production through defined LOT number system
- Well-to-well spacing: 4.5mm
- μClear™ bottom
Certifications
DIN ISO 9001
Specifications
Specifications
| Binding Property | High Binding |
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Binding Type | High Binding |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Well Shape | Flat Bottom (F-well) |