missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 380 Foam Nitrile Coated Seamless Knit Nylon Gloves

Product Code. 17806101
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17806101

Brand: SHOWA™ 38007M

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

General purpose gloves provide a long lasting grip in wet or dry conditions while also ensuring tactile precision while working with oily parts or lifting heavy objects.

  • Long-lasting grip in both wet and dry environments
  • Ensures tactile precision while working with small oily components or lifting heavy objects
  • Elasticated wrist cuff prevents debris from entering the glove
  • Open-back design allows air and moisture to escape

Specifications

Construction Seamless Knit
Coating Material Nitrile Foam
Color Blue
Hand Specific
Latex Free No
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
For Use With (Application) Automotive Repairs, and Intricate/Oily Parts Handling
Glove Cuff Style Knit
Series 380
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.