Learn More
Jenway™ 3520 pH/temp meter with glass combination pH electrode
Product Code. 11420489
Change view
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) Unit Size
11420489 -2 to 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11420489 Supplier Jenway™ Supplier No. 3545912
  • ONLINE EXCLUSIVE PRICE
    1200.00€
    1017.59€ / Pack of 1
    Save 182.41€
    15% Off
View all promotions
Estimated Shipment: 23-12-2026
3520 pH/temp meter with glass combination pH electrode, stand and holder, ATC probe, BNC shorting plug, EU power supply offers research grade specifications with a comprehensive range of features and functions, making it suitable for the broadest range of research, general laboratory, QC and GLP based applications.
  • Ideal for quality control and GLP applications
  • Up to 3 decimal place resolution
  • 1, 2 or 3 point pH calibration
  • Automatic or manual buffer selection
  • Multiple language options
  • Storage of up to 500 results
  • RS232 connection to printer or PC or infra red (IrDA) communication
  • GLP compliant
  • 3 year warranty
Specifications
Includes Glass combination pH electrode, electrode stand and holder, ATC probe, BNC shorting plug, pH 4, 7 and 10 buffer sachets and EU power supply
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±2
Accuracy (pH) ±0.003
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) ±1999.9
Resolution (mV) 0.1/1mV
Calibration Points 1, 2 or 3
Temperature Range (Metric) -10°C to +105°C
Resolution (Temperature) 0.1°C
Dimensions (L x W x H) 210 x 250 x 55 mm
Weight (Metric) 850 g
Power Supply EU
Electrical Requirements 9V AC ± 10, 50/ 60Hz