Learn More
Cole-Parmer™ 316 Stainless Steel Turbine Propeller
Product Code. 11770099 Shop All Cole-Parmer Products
Click to view available options
For Use With (Equipment):
Cole-Parmer Vortex Mixers
Unit Size:
Each
Product Code. 11770099
Brand: Cole-Parmer™ 0455215
Only null left
Add to basket
Add to basket
Ideal for airless mixing in deep containers.
  • 316 Stainless Steel Turbine Propeller
  • 2 x 3/8 in. (dia. x bore dia.)
  • Update clamps, mounts, shafts and propellers to accommodate new protocols
  • Replacement parts keep your overhead mixer running smoothly
Specifications
Product Type Turbine Propeller
Material Stainless Steel (316)
Diameter (Metric) 51 mm
For Use With (Application) Airless mixing in deep containers
For Use With (Equipment) Cole-Parmer Vortex Mixers