Learn More
Cole-Parmer™ 316 Stainless Steel Turbine Propeller
Shop All Cole-Parmer Products
Click to view available options
For Use With (Equipment):
Cole-Parmer Vortex Mixers
Unit Size:
Each
Description
- 316 Stainless Steel Turbine Propeller
- 2 x 3/8 in. (dia. x bore dia.)
- Update clamps, mounts, shafts and propellers to accommodate new protocols
- Replacement parts keep your overhead mixer running smoothly
Specifications
Specifications
| Product Type | Turbine Propeller |
| Material | Stainless Steel (316) |
| Diameter (Metric) | 51 mm |
| For Use With (Application) | Airless mixing in deep containers |
| For Use With (Equipment) | Cole-Parmer Vortex Mixers |