Learn More
Masterflex™ Elbow Fitting, NPT(M) x Barb, 316 Stainless Steel
Product Code. 11705658
Click to view available options
Tubing Size:
1/8 NPT Thread x 1/4 in. ID tubing
Unit Size:
Each
Product Code. 11705658
Brand: Masterflex™ 3120896
Only null left
Add to basket
Add to basket
Cole-Parmer™ Elbow Fitting, NPT(M) x Barb, 316 Stainless Steel is economical, easy to use, and suitable for low-pressure applications.
  • 316 stainless steel construction
  • Ideal for low-pressure applications (generally less than 30psi)
Specifications
For Use With 1/8 NPT Thread x 1/4 in. ID tubing
Packaging Quantity 1 Each
Tubing Size 1/8 NPT Thread x 1/4 in. ID tubing
Port Size 3/16 in. orifice; 23/32 x 11/16 in.
Thread Size 1/8 in. NPT