Learn More
Masterflex™ Elbow Fitting, NPT(M) x Barb, 316 Stainless Steel
Click to view available options
Tubing Size:
1/8 NPT Thread x 1/4 in. ID tubing
Unit Size:
Each
Description
- 316 stainless steel construction
- Ideal for low-pressure applications (generally less than 30psi)
Specifications
Specifications
| For Use With | 1/8 NPT Thread x 1/4 in. ID tubing |
| Packaging Quantity | 1 Each |
| Tubing Size | 1/8 NPT Thread x 1/4 in. ID tubing |
| Port Size | 3/16 in. orifice; 23/32 x 11/16 in. |
| Thread Size | 1/8 in. NPT |