Learn More
Linker Industrietechnik™ Stainless Steel Test Sieve with Soft Soldered Seam, 315 Dia. x 55 mm Height
Product Code. 11775398
Click to view available options
Pore Size:
40 μm
50 μm
75 μm
80 μm
1 mm
100 μm
125 μm
1.4 mm
1.7 mm
2 mm
200 μm
250 μm
3.15 mm
315 μm
4 mm
4.5 mm
5 mm
500 μm
5.6 mm
560 μm
10 mm
Unit Size:
Each
Product Code. 11775398
Brand: Linker Industrietechnik™ 553.155.222058
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Sifts particles in a wide range of pore sizes. These stainless-steel test sieves have soft soldered seams and an internal height of 55 mm and diameter of 315 mm.
  • Stainless-steel racks with an internal height of 55 mm
  • Diameter: 315 mm
  • Stainless-steel test sieve skin
  • Soft soldered seam
  • Identification number and certificate of compliance
  • Resistant up to temperatures of 170°C
Specifications
Quantity 1Each
Pore Size 1.4 mm
Type Test Sieve
Certifications/Compliance Compliance 2.1 EN 10 204 and ISO 3310-1. Main range in accordance with ISO 565 (mesh apertures)
Material Stainless Steel
Height (Metric) 55mm
Diameter (Metric) Outer 315 mm
Mesh Material Stainless Steel