Specifications
| Quantity | 1Each |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E 11 |
| Material | Stainless Steel |
| For Use With (Equipment) | Shaker AS 300 |
| Includes | Certificate Indicating Compliance, 7 test sieves Includes (30 Mesh, 16 Mesh, 8 Mesh, 4 Mesh, 3/8 in., 3/4 in., 1 1/4 in.) and Collecting Pan |
| Height (Metric) | 40 mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 305 mm |
| Frame Material | Stainless Steel |
| Show More |