Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Product Code. 11583254
Change view
Click to view available options
Needle Gauge:
17 G
19 G
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge unitSize
11583254 17 G Pack of 6
11738386 19 G Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11583254 Supplier Hamilton™ Supplier No. 775108
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Material Nickel-plated Brass (Hub), Stainless Steel (304)
Needle Gauge 17 G
Diameter (Metric) Inner 1.07 mm
Diameter (Metric) Outer 1.47 mm
Needle Point Style 5 or AS
For Use With (Equipment) TLL Syringes and LT or TLL Connectors