missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine Bis(T-butyl Ester) Dihydrochloride, TRC

Product Code. 30366977
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366977 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366977 Supplier Toronto Research Chemicals Supplier No. TRCC99795010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Cystine Bis(T-butyl Ester) Dihydrochloride
CAS 38261-78-8
Molecular Formula C14 H28 N2 O4 S2
Quantity 10 g
Synonym Cystine, Di-tert-butyl Ester, Dihydrochloride, L-Cystine, Bis(1,1-dimethylethyl) Ester, Dihydrochloride, L-Cystine Di-tert-butyl Ester Dihydrochloride
InChI Formula InChI=1S/C14H28N2O4S2/c1-13(2,3)19-11(17)9(15)7-21-22-8-10(16)12(18)20-14(4,5)6/h9-10H,7-8,15-16H2,1-6H3/t9-,10-/m0/s1
SMILES CC(C)(C)OC(=O)[C@@H](N)CSSC[C@H](N)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2R)-2-amino-3-[[(2R)-2-amino-3-[(2-methylpropan-2-yl)oxy]-3-oxopropyl]disulfanyl]propanoate
Molecular Weight (g/mol) 352.513
Formula Weight 352.149
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.