Learn More
KGW Isotherm™ 3 L Dewar Flask with Aluminum Jacket
Specifications
Specifications
| Diameter (Metric) Outer | 160mm |
| For Use With (Application) | Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants |
| Height (Metric) Inner | 230mm |
| Height (Metric) | 280mm |
| Capacity (Metric) | 3 L |
| Diameter (Metric) Inner | 138 mm |
| Material | Borosilicate Glass |