missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propargyl alcohol, 98%

Product Code. 11302187
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302187 25 g 25g
11382177 1 g 1g
11392177 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302187 Supplier Thermo Scientific Alfa Aesar Supplier No. L09649.14
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5272-36-6 | C6H12OSi | 128.25 g/mol

3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.

Solubility
Not miscible with water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Protect from heat. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, heat and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5272-36-6
Molecular Formula C6H12OSi
Molecular Weight (g/mol) 128.25
MDL Number MFCD00002913
InChI Key ZVGCJDPEKKEYES-UHFFFAOYSA-N
Synonym 3-trimethylsilyl-2-propyn-1-ol, 3-trimethylsilyl propargyl alcohol, 2-propyn-1-ol, 3-trimethylsilyl, 3-trimethylsilyl prop-2-yn-1-ol, 1-trimethylsilylpropargyl alcohol, acmc-209l1b, 3-trimethylsilylpropargyl alcohol, 3-trimethylsilanyl-prop-2-yn-1-ol, 3-trimethylsilyl-2-propyn-1-ol #
PubChem CID 78930
IUPAC Name 3-trimethylsilylprop-2-yn-1-ol
SMILES C[Si](C)(C)C#CCO

Specifications

Boiling Point 170°C to 172°C
Quantity 25 g
Linear Formula (CH3)3SiC≡CCH2OH
UN Number UN2810
Beilstein 1902505
Flash Point 66°C (150°F)
Solubility Information Not miscible with water.
Formula Weight 128.25
Percent Purity ≥98%
Refractive Index 1.451
Sensitivity Moisture sensitive
Density 0.865
Chemical Name or Material 3-(Trimethylsilyl)propargyl alcohol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.