Learn More
3-(Trifluoromethyl)pivalanilide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Trifluoromethyl)pivalanilide |
| CAS | 1939-19-1 |
| Molecular Formula | C12 H14 F3 N O |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C12H14F3NO/c1-11(2,3)10(17)16-9-6-4-5-8(7-9)12(13,14)15/h4-7H,1-3H3,(H,16,17) |
| SMILES | CC(C)(C)C(=O)Nc1cccc(c1)C(F)(F)F |
| IUPAC Name | 2,2-dimethyl-N-[3-(trifluoromethyl)phenyl]propanamide |
| Molecular Weight (g/mol) | 245.24 |
| Formula Weight | 245.1027 |