missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)oxetan-3-ol, TRC

Product Code. 30416410
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416410 100 mg -
30420102 500 mg 500mg
30420984 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416410 Supplier Toronto Research Chemicals Supplier No. TRCT222086100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1403676-72-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.08
IUPAC Name 3-(trifluoromethyl)oxetan-3-ol
SMILES OC1(COC1)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H5F3O2/c5-4(6,7)3(8)1-9-2-3/h8H,1-2H2
Formula Weight 142.0242
Chemical Name or Material 3-(Trifluoromethyl)oxetan-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.